The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000109.3 | NP_000100.2 | 55 - 56 | 8540 | 122 |
NM_004006.2 | NP_003997.1 | 55 - 56 | 8464 | 122 | |
NM_004009.3 | NP_004000.1 | 55 - 56 | 8471 | 122 | |
NM_004010.3 | NP_004001.1 | 55 - 56 | 8554 | 122 | |
NM_004011.3 | NP_004002.2 | 27 - 28 | 4242 | 122 | |
NM_004012.3 | NP_004003.1 | 27 - 28 | 4385 | 122 | |
NM_004013.2 | NP_004004.1 | 12 - 13 | 1881 | 122 | |
NM_004020.3 | NP_004011.2 | 12 - 13 | 1881 | 122 | |
NM_004021.2 | NP_004012.1 | 12 - 13 | 1881 | 122 | |
NM_004022.2 | NP_004013.1 | 12 - 13 | 1881 | 122 | |
NM_004023.2 | NP_004014.1 | 12 - 13 | 1881 | 122 | |
XM_006724468.2 | XP_006724531.1 | 55 - 56 | 8417 | 122 | |
XM_006724469.3 | XP_006724532.1 | 55 - 56 | 8391 | 122 | |
XM_006724470.3 | XP_006724533.1 | 55 - 56 | 8417 | 122 | |
XM_006724473.2 | XP_006724536.1 | 54 - 55 | 8279 | 122 | |
XM_006724474.3 | XP_006724537.1 | 55 - 56 | 8417 | 122 | |
XM_006724475.2 | XP_006724538.1 | 55 - 56 | 8417 | 122 | |
XM_011545467.1 | XP_011543769.1 | 54 - 55 | 8294 | 122 | |
XM_011545468.2 | XP_011543770.1 | 55 - 56 | 8417 | 122 | |
XM_017029328.1 | XP_016884817.1 | 55 - 56 | 8417 | 122 | |
XM_017029331.1 | XP_016884820.1 | 15 - 16 | 2993 | 122 | |
GenBank mRNA | AB208836.1 | - | 9 - 10 | 1328 | 122 |
BC127103.1 | - | 9 - 10 | 1405 | 122 | |
BC150141.1 | - | 9 - 10 | 1405 | 122 | |
M18533.1 | - | 55 - 56 | 8428 | 122 | |
X14298.1 | - | 55 - 56 | 8318 | 122 |