Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHS1
Entrez Gene ID
4868
Gene Name
NPHS1, nephrin
Gene Aliases
CNF, NPHN, nephrin
Chromosome Location
Chr.19: 35825372 - 35851993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004646.3 NP_004637.1 11 - 12 1600 91
GenBank mRNA AF035835.1 - 11 - 12 1444 91
AF190637.1 - 11 - 12 1444 91
EU642886.1 - 11 - 12 1444 91
EU642887.1 - 11 - 12 1443 91

View Full Product Details