Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
Chromosome Location
Chr.5: 180300690 - 180353387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005110.3 NP_005101.1 12 - 13 1322 74
XM_006714942.2 XP_006715005.1 12 - 13 1271 74
XM_017010100.1 XP_016865589.1 11 - 12 1207 74
GenBank mRNA AB016789.1 - 12 - 13 1251 74
AK001242.1 - 2 - 3 204 74
AK022507.1 - 12 - 13 1262 74
AK297750.1 - 10 - 11 958 74
AK314551.1 - 12 - 13 1262 74
BC000012.2 - 12 - 13 1260 74
BT009818.1 - 11 - 12 1152 74
KU178494.1 - 11 - 12 1152 74
KU178495.1 - 11 - 12 1152 74

View Full Product Details