Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
Chromosome Location
Chr.5: 180300690 - 180353387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005110.3 NP_005101.1 4 - 5 507 97
XM_006714942.2 XP_006715005.1 4 - 5 456 97
XM_017010100.1 XP_016865589.1 4 - 5 454 97
GenBank mRNA AB016789.1 - 4 - 5 436 97
AK022507.1 - 4 - 5 447 97
AK297750.1 - 2 - 3 143 97
AK314551.1 - 4 - 5 447 97
BC000012.2 - 4 - 5 445 97
BT009818.1 - 3 - 4 337 97
KU178494.1 - 3 - 4 337 97
KU178495.1 - 3 - 4 337 97

View Full Product Details