Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
Chromosome Location
Chr.2: 74557640 - 74648338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281295.1 NP_001268224.1 1 - 2 77 117
NM_001281296.1 NP_001268225.1 1 - 2 77 117
NM_001321739.1 NP_001308668.1 1 - 2 77 117
GenBank mRNA AK300188.1 - 1 - 2 69 117
DR158220.1 - 1 - 2 77 117

View Full Product Details