Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAN
Entrez Gene ID
176
Gene Name
aggrecan
Gene Aliases
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK
Chromosome Location
Chr.15: 88803442 - 88875354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135.3 NP_001126.3 15 - 16 7589 61
GenBank mRNA M55172.1 - 16 - 17 6930 61

View Full Product Details