The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100878.1 | NP_001094348.1 | 14 - 14 | 3200 | 103 |
XM_011517214.1 | XP_011515516.1 | 14 - 14 | 2570 | 103 | |
XM_011517215.1 | XP_011515517.1 | 14 - 14 | 2567 | 103 | |
XM_011517216.2 | XP_011515518.1 | 13 - 13 | 2502 | 103 | |
XM_011517217.1 | XP_011515519.1 | 12 - 12 | 2436 | 103 | |
XM_011517218.1 | XP_011515520.1 | 13 - 13 | 3077 | 103 | |
XM_011517221.2 | XP_011515523.1 | 15 - 15 | 2335 | 103 | |
XM_011517222.1 | XP_011515524.1 | 14 - 14 | 2267 | 103 | |
XM_011517223.1 | XP_011515525.1 | 13 - 13 | 2201 | 103 | |
GenBank mRNA | AF086255.1 | - | 1 - 1 | 419 | 103 |
AF289596.1 | - | 3 - 3 | 2262 | 103 | |
BC029031.1 | - | 5 - 5 | 1870 | 103 |