Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHC1
Entrez Gene ID
6464
Gene Name
SHC adaptor protein 1
Gene Aliases
SHC, SHCA
Chromosome Location
Chr.1: 154962298 - 154974492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130040.1 NP_001123512.1 7 - 8 1206 57
NM_001130041.1 NP_001123513.1 8 - 9 894 57
NM_001202859.1 NP_001189788.1 8 - 9 894 57
NM_003029.4 NP_003020.2 8 - 9 894 57
NM_183001.4 NP_892113.4 7 - 8 1206 57
XM_005245449.4 XP_005245506.1 7 - 8 2422 57
XM_005245451.4 XP_005245508.1 8 - 9 817 57
XM_011509898.2 XP_011508200.1 7 - 8 735 57
XM_017002081.1 XP_016857570.1 6 - 7 2364 57
XM_017002082.1 XP_016857571.1 6 - 7 2365 57
XM_017002083.1 XP_016857572.1 7 - 8 804 57
GenBank mRNA AB208849.1 - 4 - 5 323 57
AB451255.1 - 7 - 8 985 57
AB451379.1 - 7 - 8 985 57
AK096169.1 - 7 - 8 1189 57
AK130014.1 - 6 - 7 539 57
AK225332.1 - 7 - 8 1131 57
AK292143.1 - 8 - 9 894 57
AK296786.1 - 8 - 9 1017 57
AK315842.1 - 8 - 9 805 57
BC014158.2 - 8 - 9 775 57
BC033925.1 - 5 - 6 608 57
BX647136.1 - 8 - 9 1122 57
BX647149.1 - 7 - 8 1206 57
U73377.1 - 7 - 8 1179 57
X68148.1 - 8 - 9 736 57

View Full Product Details