The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025250.2 | NP_079526.1 | 7 - 8 | 1056 | 76 |
XM_011515535.2 | XP_011513837.1 | 7 - 8 | 1024 | 76 | |
XM_017012655.1 | XP_016868144.1 | 6 - 7 | 930 | 76 | |
XM_017012656.1 | XP_016868145.1 | 6 - 7 | 928 | 76 | |
GenBank mRNA | AB051478.2 | - | 7 - 8 | 1051 | 76 |
AB162931.1 | - | 7 - 8 | 881 | 76 | |
AK074158.1 | - | 3 - 4 | 364 | 76 | |
AK124608.1 | - | 3 - 4 | 499 | 76 | |
BC131824.1 | - | 7 - 8 | 927 | 76 | |
BC152447.1 | - | 7 - 8 | 1051 | 76 |