The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025250.2 | NP_079526.1 | 2 - 3 | 483 | 72 |
XM_011515535.2 | XP_011513837.1 | 2 - 3 | 451 | 72 | |
XM_017012655.1 | XP_016868144.1 | 2 - 3 | 453 | 72 | |
XM_017012656.1 | XP_016868145.1 | 2 - 3 | 451 | 72 | |
GenBank mRNA | AB051478.2 | - | 2 - 3 | 478 | 72 |
AB162931.1 | - | 2 - 3 | 308 | 72 | |
AL522452.3 | - | 2 - 3 | 481 | 72 | |
BC131824.1 | - | 2 - 3 | 354 | 72 | |
BC152447.1 | - | 2 - 3 | 478 | 72 |