The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014215.2 | NP_055030.1 | 1 - 2 | 390 | 63 |
GenBank mRNA | AK310760.1 | - | 1 - 2 | 391 | 63 |