The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000528.3 | NP_000519.2 | 11 - 12 | 1495 | 90 |
NM_001173498.1 | NP_001166969.1 | 11 - 12 | 1492 | 90 | |
XM_005259913.1 | XP_005259970.1 | 11 - 12 | 1476 | 90 | |
XM_017026818.1 | XP_016882307.1 | 8 - 9 | 1074 | 90 | |
GenBank mRNA | AB209921.1 | - | 11 - 12 | 1407 | 90 |
AK291572.1 | - | 11 - 12 | 1460 | 90 | |
AK303418.1 | - | 12 - 13 | 1434 | 90 | |
BC000736.2 | - | 11 - 12 | 1441 | 90 | |
U05572.1 | - | 11 - 12 | 1449 | 90 | |
U60266.1 | - | 11 - 12 | 1433 | 90 | |
U68382.1 | - | 10 - 11 | 1317 | 90 | |
U68567.1 | - | 11 - 12 | 1407 | 90 |