The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270488.1 | NP_001257417.1 | 2 - 3 | 434 | 139 |
NM_005643.3 | NP_005634.1 | 2 - 3 | 434 | 139 | |
XM_011514827.2 | XP_011513129.1 | 2 - 3 | 824 | 139 | |
GenBank mRNA | AF118094.1 | - | 2 - 3 | 174 | 139 |
AK302226.1 | - | 2 - 3 | 405 | 139 | |
AK313475.1 | - | 2 - 3 | 375 | 139 | |
BC021972.2 | - | 2 - 3 | 390 | 139 | |
BT019404.1 | - | 2 - 3 | 320 | 139 | |
BT019405.1 | - | 2 - 3 | 320 | 139 | |
BX538195.1 | - | 1 - 2 | 231 | 139 | |
CR542068.1 | - | 2 - 3 | 320 | 139 | |
D63705.1 | - | 2 - 3 | 403 | 139 | |
DA153455.1 | - | 2 - 3 | 434 | 139 | |
X83928.1 | - | 2 - 3 | 412 | 139 |