Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIMKLB
Entrez Gene ID
57494
Gene Name
ribosomal modification protein rimK like family member B
Gene Aliases
FAM80B, NAAGS, NAAGS-I
Chromosome Location
Chr.12: 8681606 - 8783098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297776.1 NP_001284705.1 5 - 6 1151 68
NM_020734.3 NP_065785.2 5 - 6 1583 68
NR_123740.1 5 - 6 895 68
XM_006719118.2 XP_006719181.1 5 - 6 1272 68
XM_006719127.2 XP_006719190.1 5 - 6 1272 68
XM_011520776.1 XP_011519078.1 5 - 6 1272 68
XM_011520777.1 XP_011519079.1 5 - 6 1272 68
XM_011520778.2 XP_011519080.1 5 - 6 1272 68
XM_011520779.2 XP_011519081.1 5 - 6 1272 68
XM_011520780.1 XP_011519082.1 5 - 6 1272 68
XM_011520783.1 XP_011519085.1 5 - 6 1272 68
XM_011520790.1 XP_011519092.1 6 - 7 1959 68
XM_017019677.1 XP_016875166.1 5 - 6 1272 68
XM_017019678.1 XP_016875167.1 5 - 6 1272 68
XM_017019679.1 XP_016875168.1 5 - 6 1272 68
XM_017019680.1 XP_016875169.1 5 - 6 1272 68
XM_017019681.1 XP_016875170.1 5 - 6 1272 68
XM_017019682.1 XP_016875171.1 5 - 6 1151 68
XM_017019683.1 XP_016875172.1 6 - 7 1959 68
XM_017019684.1 XP_016875173.1 5 - 6 895 68
XM_017019685.1 XP_016875174.1 6 - 7 1674 68
XM_017019686.1 XP_016875175.1 5 - 6 909 68
XM_017019687.1 XP_016875176.1 6 - 7 1273 68
XM_017019688.1 XP_016875177.1 5 - 6 1413 68
XM_017019689.1 XP_016875178.1 5 - 6 962 68
XM_017019690.1 XP_016875179.1 5 - 6 773 68
XM_017019691.1 XP_016875180.1 5 - 6 1151 68
XM_017019692.1 XP_016875181.1 5 - 6 1129 68
XM_017019693.1 XP_016875182.1 5 - 6 1151 68
XM_017019694.1 XP_016875183.1 5 - 6 1156 68
XM_017019695.1 XP_016875184.1 5 - 6 925 68
XM_017019696.1 XP_016875185.1 6 - 7 1959 68
XM_017019697.1 XP_016875186.1 5 - 6 846 68
GenBank mRNA AB033064.1 - 5 - 6 1522 68
AK302847.1 - 5 - 6 1151 68
BC006368.2 - 6 - 7 1844 68
BC021977.2 - 2 - 3 247 68
BC033793.1 - 5 - 6 892 68

View Full Product Details