The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_123740.1 | 6 - 7 | 1036 | 149 | |
XM_006719127.2 | XP_006719190.1 | 6 - 7 | 1413 | 149 | |
XM_011520780.1 | XP_011519082.1 | 6 - 7 | 1413 | 149 | |
XM_011520783.1 | XP_011519085.1 | 6 - 7 | 1413 | 149 | |
XM_011520790.1 | XP_011519092.1 | 7 - 8 | 2100 | 149 | |
XM_017019680.1 | XP_016875169.1 | 6 - 7 | 1413 | 149 | |
XM_017019691.1 | XP_016875180.1 | 6 - 7 | 1292 | 149 | |
XM_017019692.1 | XP_016875181.1 | 6 - 7 | 1270 | 149 | |
XM_017019693.1 | XP_016875182.1 | 6 - 7 | 1292 | 149 | |
XM_017019694.1 | XP_016875183.1 | 6 - 7 | 1297 | 149 | |
XM_017019695.1 | XP_016875184.1 | 6 - 7 | 1066 | 149 | |
XM_017019696.1 | XP_016875185.1 | 7 - 8 | 2100 | 149 | |
XM_017019697.1 | XP_016875186.1 | 6 - 7 | 987 | 149 | |
GenBank mRNA | BC033793.1 | - | 6 - 7 | 1033 | 149 |