Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNE2
Entrez Gene ID
9134
Gene Name
cyclin E2
Gene Aliases
CYCE2
Chromosome Location
Chr.8: 94880224 - 94896671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057749.2 NP_477097.1 4 - 5 274 96
XM_011517366.2 XP_011515668.1 3 - 4 599 96
XM_017013957.1 XP_016869446.1 3 - 4 599 96
XM_017013958.1 XP_016869447.1 4 - 5 416 96
XM_017013959.1 XP_016869448.1 4 - 5 518 96
GenBank mRNA AF091433.1 - 3 - 4 207 96
AF102778.1 - 2 - 3 170 96
AF106690.1 - 2 - 3 170 96
AF112857.1 - 2 - 3 170 96
BC020729.1 - 4 - 5 221 96
BG720611.1 - 4 - 5 279 96
BJ990268.1 - 4 - 5 274 96

View Full Product Details