Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ABCC13
- Entrez Gene ID
-
150000
- Gene Name
- ATP binding cassette subfamily C member 13 (pseudogene)
- Gene Aliases
-
ABCC13P, C21orf73, PRED6
- Chromosome Location
-
Chr.21: 14273799 - 14301386 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF418600.1 |
- |
5 - 6 |
889 |
130 |
View Full Product Details