The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000599.3 | NP_000590.1 | 2 - 3 | 1344 | 123 |
GenBank mRNA | AF055033.1 | - | 2 - 3 | 1321 | 123 |
AK314992.1 | - | 2 - 3 | 622 | 123 | |
BC011453.1 | - | 2 - 3 | 918 | 123 | |
BT019706.1 | - | 2 - 3 | 570 | 123 | |
BT019707.1 | - | 2 - 3 | 570 | 123 | |
CR456809.1 | - | 2 - 3 | 570 | 123 | |
M62782.1 | - | 2 - 3 | 626 | 123 | |
M65062.1 | - | 2 - 3 | 613 | 123 |