The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287395.1 | NP_001274324.1 | 15 - 16 | 1811 | 57 |
XM_011511075.2 | XP_011509377.1 | 15 - 16 | 1988 | 57 | |
XM_011511076.2 | XP_011509378.1 | 15 - 16 | 1988 | 57 | |
XM_011511077.2 | XP_011509379.1 | 15 - 16 | 1988 | 57 | |
XM_011511078.2 | XP_011509380.1 | 15 - 16 | 1988 | 57 | |
XM_011511079.2 | XP_011509381.1 | 15 - 16 | 1988 | 57 | |
XM_011511080.2 | XP_011509382.1 | 15 - 16 | 3459 | 57 | |
XM_011511081.2 | XP_011509383.1 | 15 - 16 | 1988 | 57 | |
XM_011511082.2 | XP_011509384.1 | 15 - 16 | 1988 | 57 | |
XM_011511083.2 | XP_011509385.1 | 15 - 16 | 1988 | 57 | |
XM_011511084.2 | XP_011509386.1 | 15 - 16 | 1988 | 57 | |
XM_011511085.2 | XP_011509387.1 | 9 - 10 | 1058 | 57 | |
XM_011511086.2 | XP_011509388.1 | 15 - 16 | 1988 | 57 |