The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025045.5 | NP_079321.3 | 7 - 8 | 755 | 56 |
XM_005261751.4 | XP_005261808.1 | 8 - 9 | 756 | 56 | |
XM_011530379.2 | XP_011528681.1 | 7 - 8 | 872 | 56 | |
XM_011530380.2 | XP_011528682.1 | 6 - 7 | 658 | 56 | |
XM_011530381.2 | XP_011528683.1 | 7 - 8 | 649 | 56 | |
XM_011530382.2 | XP_011528684.1 | 7 - 8 | 1037 | 56 | |
XM_011530383.2 | XP_011528685.1 | 6 - 7 | 975 | 56 | |
XM_011530384.2 | XP_011528686.1 | 7 - 8 | 913 | 56 | |
GenBank mRNA | AY358164.1 | - | 7 - 8 | 755 | 56 |