The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012174.1 | NP_036306.1 | 4 - 5 | 563 | 111 |
NM_153348.2 | NP_699179.2 | 4 - 5 | 761 | 111 | |
XM_017019175.1 | XP_016874664.1 | 4 - 5 | 538 | 111 | |
XM_017019176.1 | XP_016874665.1 | 5 - 6 | 770 | 111 | |
XM_017019177.1 | XP_016874666.1 | 5 - 6 | 770 | 111 | |
GenBank mRNA | AF176707.1 | - | 4 - 5 | 541 | 111 |
AK289621.1 | - | 4 - 5 | 554 | 111 | |
BC037296.2 | - | 4 - 5 | 761 | 111 |