The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002019.4 | NP_002010.2 | 19 - 20 | 2995 | 92 |
GenBank mRNA | AB209050.1 | - | 3 - 4 | 351 | 92 |
AF063657.2 | - | 19 - 20 | 2710 | 92 | |
DQ836394.1 | - | 5 - 6 | 535 | 92 | |
DQ836395.1 | - | 6 - 7 | 597 | 92 | |
EF491868.1 | - | 1 - 2 | 244 | 92 |