The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003106.3 | NP_003097.1 | 1 - 1 | 1539 | 91 |
GenBank mRNA | AW016610.1 | - | 1 - 1 | 310 | 91 |