The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134830.1 | NP_001128302.1 | 14 - 15 | 2498 | 102 |
NM_001134831.1 | NP_001128303.1 | 16 - 17 | 2645 | 102 | |
NM_001134832.1 | NP_001128304.1 | 15 - 16 | 2560 | 102 | |
NM_017651.4 | NP_060121.3 | 15 - 16 | 2560 | 102 | |
XM_011535910.2 | XP_011534212.1 | 17 - 18 | 2789 | 102 | |
XM_011535911.2 | XP_011534213.1 | 15 - 16 | 2519 | 102 | |
XM_017010977.1 | XP_016866466.1 | 16 - 17 | 2764 | 102 | |
XM_017010978.1 | XP_016866467.1 | 15 - 16 | 2462 | 102 | |
XM_017010979.1 | XP_016866468.1 | 15 - 16 | 2462 | 102 | |
XM_017010980.1 | XP_016866469.1 | 15 - 16 | 2462 | 102 | |
XM_017010981.1 | XP_016866470.1 | 15 - 16 | 2597 | 102 | |
XM_017010982.1 | XP_016866471.1 | 15 - 16 | 2462 | 102 | |
XM_017010983.1 | XP_016866472.1 | 15 - 16 | 2462 | 102 | |
XM_017010984.1 | XP_016866473.1 | 15 - 16 | 2462 | 102 | |
GenBank mRNA | AJ459824.1 | - | 15 - 16 | 2540 | 102 |
AJ459825.1 | - | 15 - 16 | 2540 | 102 | |
AJ606362.1 | - | 14 - 15 | 2478 | 102 | |
AK000076.1 | - | 7 - 8 | 961 | 102 | |
AK024085.1 | - | 3 - 4 | 453 | 102 | |
AK092262.1 | - | 14 - 15 | 2471 | 102 | |
AK225210.1 | - | 7 - 8 | 961 | 102 | |
AL136797.1 | - | 13 - 14 | 2279 | 102 | |
BC015019.1 | - | 5 - 6 | 760 | 102 | |
BC094800.1 | - | 16 - 17 | 2568 | 102 | |
DQ090887.1 | - | 16 - 17 | 2599 | 102 |