The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134830.1 | NP_001128302.1 | 7 - 8 | 1391 | 153 |
NM_001134831.1 | NP_001128303.1 | 9 - 10 | 1538 | 153 | |
NM_001134832.1 | NP_001128304.1 | 8 - 9 | 1453 | 153 | |
NM_017651.4 | NP_060121.3 | 8 - 9 | 1453 | 153 | |
XM_011535910.2 | XP_011534212.1 | 10 - 11 | 1682 | 153 | |
XM_011535911.2 | XP_011534213.1 | 8 - 9 | 1412 | 153 | |
XM_017010977.1 | XP_016866466.1 | 9 - 10 | 1657 | 153 | |
XM_017010978.1 | XP_016866467.1 | 8 - 9 | 1355 | 153 | |
XM_017010979.1 | XP_016866468.1 | 8 - 9 | 1355 | 153 | |
XM_017010980.1 | XP_016866469.1 | 8 - 9 | 1355 | 153 | |
XM_017010981.1 | XP_016866470.1 | 8 - 9 | 1490 | 153 | |
XM_017010982.1 | XP_016866471.1 | 8 - 9 | 1355 | 153 | |
XM_017010983.1 | XP_016866472.1 | 8 - 9 | 1355 | 153 | |
XM_017010984.1 | XP_016866473.1 | 8 - 9 | 1355 | 153 | |
GenBank mRNA | AJ459824.1 | - | 8 - 9 | 1433 | 153 |
AJ459825.1 | - | 8 - 9 | 1433 | 153 | |
AJ606362.1 | - | 7 - 8 | 1371 | 153 | |
AK092262.1 | - | 7 - 8 | 1364 | 153 | |
AL136797.1 | - | 6 - 7 | 1172 | 153 | |
BC065712.1 | - | 6 - 7 | 1217 | 153 | |
BC094800.1 | - | 8 - 9 | 1428 | 153 | |
DQ090887.1 | - | 9 - 10 | 1492 | 153 |