Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
Chromosome Location
Chr.1: 39084167 - 39487138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012090.5 NP_036222.3 26 - 27 3518 104
XM_005270694.1 XP_005270751.1 22 - 23 4179 104
XM_005270695.1 XP_005270752.1 22 - 23 4179 104
XM_005270696.1 XP_005270753.1 22 - 23 4179 104
XM_005270697.1 XP_005270754.1 22 - 23 4179 104
XM_005270698.1 XP_005270755.1 22 - 23 4179 104
XM_006710519.1 XP_006710582.1 22 - 23 4179 104
XM_006710520.2 XP_006710583.1 22 - 23 4179 104
XM_006710521.1 XP_006710584.1 22 - 23 4179 104
XM_006710522.1 XP_006710585.1 22 - 23 4179 104
XM_006710524.2 XP_006710587.1 22 - 23 4179 104
XM_006710526.3 XP_006710589.1 27 - 28 3611 104
XM_006710528.3 XP_006710591.1 27 - 28 4172 104
XM_006710529.3 XP_006710592.1 27 - 28 3529 104
XM_006710530.3 XP_006710593.1 27 - 28 4417 104
XM_006710531.3 XP_006710594.1 27 - 28 4148 104
XM_006710532.3 XP_006710595.1 26 - 27 4076 104
XM_006710533.2 XP_006710596.1 26 - 27 4076 104
XM_006710535.3 XP_006710598.1 27 - 28 3681 104
XM_006710536.2 XP_006710599.1 22 - 23 4179 104
XM_006710538.2 XP_006710601.1 22 - 23 4179 104
XM_011541141.2 XP_011539443.1 22 - 23 4179 104
XM_011541142.2 XP_011539444.1 22 - 23 4179 104
XM_017000873.1 XP_016856362.1 22 - 23 4179 104
XM_017000874.1 XP_016856363.1 22 - 23 4179 104
XM_017000875.1 XP_016856364.1 26 - 27 3501 104
XM_017000876.1 XP_016856365.1 26 - 27 3585 104
XM_017000877.1 XP_016856366.1 27 - 28 3507 104
XM_017000878.1 XP_016856367.1 27 - 28 3370 104
XM_017000880.1 XP_016856369.1 22 - 23 4179 104
XM_017000881.1 XP_016856370.1 26 - 27 3493 104
XM_017000882.1 XP_016856371.1 22 - 23 4179 104
GenBank mRNA AB007934.3 - 6 - 7 866 104
AB029290.1 - 26 - 27 3517 104
AB033077.1 - 6 - 7 866 104
AK127519.1 - 22 - 23 4030 104

View Full Product Details