The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012090.5 | NP_036222.3 | 3 - 4 | 428 | 70 |
XM_006710532.3 | XP_006710595.1 | 3 - 4 | 986 | 70 | |
XM_006710533.2 | XP_006710596.1 | 3 - 4 | 986 | 70 | |
XM_017000875.1 | XP_016856364.1 | 3 - 4 | 411 | 70 | |
XM_017000876.1 | XP_016856365.1 | 3 - 4 | 495 | 70 | |
XM_017000881.1 | XP_016856370.1 | 3 - 4 | 403 | 70 | |
GenBank mRNA | AB029290.1 | - | 3 - 4 | 427 | 70 |
AK304414.1 | - | 3 - 4 | 537 | 70 |