Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
Chromosome Location
Chr.1: 39084167 - 39487138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012090.5 NP_036222.3 77 - 78 13850 90
XM_005270694.1 XP_005270751.1 78 - 79 20712 90
XM_005270695.1 XP_005270752.1 78 - 79 20712 90
XM_005270696.1 XP_005270753.1 78 - 79 20712 90
XM_005270697.1 XP_005270754.1 78 - 79 20712 90
XM_005270698.1 XP_005270755.1 78 - 79 20712 90
XM_006710519.1 XP_006710582.1 77 - 78 20703 90
XM_006710520.2 XP_006710583.1 77 - 78 20649 90
XM_006710521.1 XP_006710584.1 78 - 79 20712 90
XM_006710522.1 XP_006710585.1 78 - 79 20712 90
XM_006710524.2 XP_006710587.1 77 - 78 20478 90
XM_006710526.3 XP_006710589.1 83 - 84 20144 90
XM_006710528.3 XP_006710591.1 83 - 84 20705 90
XM_006710529.3 XP_006710592.1 83 - 84 20062 90
XM_006710530.3 XP_006710593.1 83 - 84 20950 90
XM_006710531.3 XP_006710594.1 83 - 84 20681 90
XM_006710532.3 XP_006710595.1 82 - 83 20609 90
XM_006710533.2 XP_006710596.1 82 - 83 20609 90
XM_006710535.3 XP_006710598.1 83 - 84 20214 90
XM_006710536.2 XP_006710599.1 73 - 74 14511 90
XM_011541141.2 XP_011539443.1 78 - 79 20658 90
XM_011541142.2 XP_011539444.1 77 - 78 15261 90
XM_017000873.1 XP_016856362.1 77 - 78 20703 90
XM_017000874.1 XP_016856363.1 77 - 78 20703 90
XM_017000875.1 XP_016856364.1 82 - 83 20034 90
XM_017000876.1 XP_016856365.1 82 - 83 20118 90
XM_017000877.1 XP_016856366.1 83 - 84 20040 90
XM_017000878.1 XP_016856367.1 83 - 84 19903 90
XM_017000879.1 XP_016856368.1 48 - 49 15474 90
XM_017000880.1 XP_016856369.1 72 - 73 14448 90
XM_017000881.1 XP_016856370.1 77 - 78 13825 90
XM_017000882.1 XP_016856371.1 72 - 73 14448 90
GenBank mRNA AB007934.3 - 55 - 56 10808 90
AB029290.1 - 77 - 78 13849 90
AF317696.1 - 47 - 48 15424 90

View Full Product Details