Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
Chromosome Location
Chr.1: 39084167 - 39487138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012090.5 NP_036222.3 79 - 80 14135 59
XM_005270694.1 XP_005270751.1 80 - 81 20997 59
XM_005270695.1 XP_005270752.1 80 - 81 20997 59
XM_005270696.1 XP_005270753.1 80 - 81 20997 59
XM_005270697.1 XP_005270754.1 80 - 81 20997 59
XM_005270698.1 XP_005270755.1 80 - 81 20997 59
XM_006710519.1 XP_006710582.1 79 - 80 20988 59
XM_006710520.2 XP_006710583.1 79 - 80 20934 59
XM_006710521.1 XP_006710584.1 80 - 81 20997 59
XM_006710522.1 XP_006710585.1 80 - 81 20997 59
XM_006710524.2 XP_006710587.1 79 - 80 20763 59
XM_006710526.3 XP_006710589.1 85 - 86 20429 59
XM_006710528.3 XP_006710591.1 85 - 86 20990 59
XM_006710529.3 XP_006710592.1 85 - 86 20347 59
XM_006710530.3 XP_006710593.1 85 - 86 21235 59
XM_006710531.3 XP_006710594.1 85 - 86 20966 59
XM_006710532.3 XP_006710595.1 84 - 85 20894 59
XM_006710533.2 XP_006710596.1 84 - 85 20894 59
XM_006710535.3 XP_006710598.1 85 - 86 20499 59
XM_006710536.2 XP_006710599.1 75 - 76 14796 59
XM_011541141.2 XP_011539443.1 80 - 81 20943 59
XM_011541142.2 XP_011539444.1 79 - 80 15546 59
XM_017000873.1 XP_016856362.1 79 - 80 20988 59
XM_017000874.1 XP_016856363.1 79 - 80 20988 59
XM_017000875.1 XP_016856364.1 84 - 85 20319 59
XM_017000876.1 XP_016856365.1 84 - 85 20403 59
XM_017000877.1 XP_016856366.1 85 - 86 20325 59
XM_017000878.1 XP_016856367.1 85 - 86 20188 59
XM_017000879.1 XP_016856368.1 50 - 51 15759 59
XM_017000880.1 XP_016856369.1 74 - 75 14733 59
XM_017000881.1 XP_016856370.1 79 - 80 14110 59
XM_017000882.1 XP_016856371.1 74 - 75 14733 59
GenBank mRNA AB007934.3 - 57 - 58 11093 59
AB029290.1 - 79 - 80 14134 59
AF317696.1 - 49 - 50 15709 59

View Full Product Details