The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013338.4 | NP_037470.1 | 2 - 3 | 304 | 97 |
GenBank mRNA | AF102850.1 | - | 2 - 3 | 312 | 97 |
AF161498.1 | - | 2 - 3 | 253 | 97 | |
AF183413.1 | - | 2 - 3 | 263 | 97 | |
AK299126.1 | - | 2 - 3 | 273 | 97 | |
BC012531.1 | - | 2 - 3 | 256 | 97 | |
BG482604.1 | - | 2 - 3 | 289 | 97 | |
BP246765.1 | - | 2 - 3 | 304 | 97 | |
CB988638.1 | - | - | 301 | 97 |