Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf42
Entrez Gene ID
374470
Gene Name
chromosome 12 open reading frame 42
Gene Aliases
-
Chromosome Location
Chr.12: 103047669 - 103525258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103526.1 5 - 6 448 72
GenBank mRNA BC039352.1 - 5 - 6 406 72

View Full Product Details