The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017964.1 | NP_001017964.1 | 3 - 4 | 457 | 111 |
GenBank mRNA | BC096754.1 | - | 3 - 4 | 530 | 111 |
BC110799.1 | - | 3 - 4 | 501 | 111 | |
BC133043.1 | - | 3 - 4 | 611 | 111 | |
BC133047.1 | - | 3 - 4 | 611 | 111 | |
BI597711.1 | - | 3 - 4 | 535 | 111 |