Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
Chromosome Location
Chr.1: 225982702 - 225999365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152608.3 NP_689821.3 1 - 2 172 85
GenBank mRNA AK092701.1 - 1 - 2 129 85
AK291008.1 - 1 - 2 135 85
AK308441.1 - 1 - 2 149 85
BC071563.1 - 1 - 2 150 85
BX648319.1 - 1 - 2 130 85
CB140971.1 - 1 - 2 172 85

View Full Product Details