Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMB1
Entrez Gene ID
3912
Gene Name
laminin subunit beta 1
Gene Aliases
CLM, LIS5
Chromosome Location
Chr.7: 107923799 - 108003359 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002291.2 NP_002282.2 33 - 34 5554 153
XM_017012201.1 XP_016867690.1 33 - 34 6022 153
GenBank mRNA BC026018.2 - 16 - 17 3112 153
BC113455.1 - 32 - 33 5262 153
BX648251.1 - 32 - 33 5483 153
M61916.1 - 33 - 34 5336 153

View Full Product Details