Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSME4
Entrez Gene ID
23198
Gene Name
proteasome activator subunit 4
Gene Aliases
PA200
Chromosome Location
Chr.2: 53864067 - 53970840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014614.2 NP_055429.2 22 - 23 2683 138
XM_006711969.1 XP_006712032.1 21 - 22 2260 138
XM_011532705.2 XP_011531007.1 23 - 24 2707 138
XM_011532706.2 XP_011531008.1 23 - 24 2707 138
XM_011532707.2 XP_011531009.1 22 - 23 2641 138
XM_011532709.2 XP_011531011.1 23 - 24 2707 138
GenBank mRNA AB621805.1 - 21 - 22 2800 138
AY894754.1 - 21 - 22 2532 138
AY894756.1 - 22 - 23 2627 138
BC112169.1 - 21 - 22 2319 138
BC113668.1 - 21 - 22 2319 138
BC143739.1 - 21 - 22 2319 138
D38521.1 - 22 - 23 2493 138

View Full Product Details