The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014614.2 | NP_055429.2 | 39 - 40 | 4580 | 126 |
XM_006711969.1 | XP_006712032.1 | 38 - 39 | 4157 | 126 | |
XM_011532705.2 | XP_011531007.1 | 40 - 41 | 4604 | 126 | |
XM_011532706.2 | XP_011531008.1 | 40 - 41 | 4604 | 126 | |
XM_011532707.2 | XP_011531009.1 | 39 - 40 | 4538 | 126 | |
GenBank mRNA | AK025517.1 | - | 8 - 9 | 924 | 126 |
AK026085.1 | - | 13 - 14 | 1577 | 126 | |
AY894754.1 | - | 38 - 39 | 4429 | 126 | |
AY894755.1 | - | 20 - 21 | 2268 | 126 | |
BC043602.1 | - | 4 - 5 | 374 | 126 | |
BC112169.1 | - | 38 - 39 | 4216 | 126 | |
BC113668.1 | - | 38 - 39 | 4216 | 126 | |
BC143739.1 | - | 38 - 39 | 4216 | 126 | |
D38521.1 | - | 39 - 40 | 4390 | 126 |