The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128852.1 | NP_001122324.1 | 19 - 20 | 2822 | 81 |
NM_001128853.1 | NP_001122325.1 | 19 - 20 | 2813 | 81 | |
NM_001128854.1 | NP_001122326.1 | 19 - 20 | 2810 | 81 | |
NM_015908.5 | NP_056992.4 | 19 - 20 | 2825 | 81 | |
XM_005250405.2 | XP_005250462.1 | 19 - 20 | 2682 | 81 | |
XM_005250406.2 | XP_005250463.1 | 19 - 20 | 2679 | 81 | |
XM_005250407.2 | XP_005250464.1 | 19 - 20 | 2670 | 81 | |
XM_005250408.2 | XP_005250465.1 | 19 - 20 | 2667 | 81 | |
XM_017012290.1 | XP_016867779.1 | 20 - 21 | 2995 | 81 | |
XM_017012291.1 | XP_016867780.1 | 20 - 21 | 2992 | 81 | |
GenBank mRNA | AF082871.1 | - | 9 - 10 | 1221 | 81 |
AF248955.1 | - | 19 - 20 | 2776 | 81 | |
AK296131.1 | - | 13 - 14 | 1791 | 81 | |
AK302820.1 | - | 19 - 20 | 2714 | 81 | |
BC000082.2 | - | 10 - 11 | 1346 | 81 | |
BC069249.1 | - | 17 - 18 | 2366 | 81 | |
BC109117.1 | - | 19 - 20 | 2704 | 81 |