Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRT
Entrez Gene ID
51593
Gene Name
serrate, RNA effector molecule
Gene Aliases
ARS2, ASR2, serrate
Chromosome Location
Chr.7: 100874971 - 100888664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128852.1 NP_001122324.1 3 - 4 518 112
NM_001128853.1 NP_001122325.1 3 - 4 518 112
NM_001128854.1 NP_001122326.1 3 - 4 518 112
NM_015908.5 NP_056992.4 3 - 4 518 112
XM_005250405.2 XP_005250462.1 3 - 4 375 112
XM_005250406.2 XP_005250463.1 3 - 4 375 112
XM_005250407.2 XP_005250464.1 3 - 4 375 112
XM_005250408.2 XP_005250465.1 3 - 4 375 112
XM_017012290.1 XP_016867779.1 4 - 5 700 112
XM_017012291.1 XP_016867780.1 4 - 5 700 112
GenBank mRNA AF248955.1 - 3 - 4 480 112
AK302820.1 - 3 - 4 518 112
AL096723.1 - 3 - 4 472 112
BC109117.1 - 3 - 4 412 112
BE297447.1 - 3 - 4 504 112
BF515651.1 - 3 - 4 355 112
BG685713.1 - 3 - 4 484 112

View Full Product Details