Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A40
Entrez Gene ID
55972
Gene Name
solute carrier family 25 member 40
Gene Aliases
MCFP
Chromosome Location
Chr.7: 87834499 - 87876377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018843.3 NP_061331.2 4 - 5 534 119
GenBank mRNA AF125531.1 - 5 - 6 507 119
AK290848.1 - 4 - 5 309 119
BC027322.1 - 3 - 4 266 119
BU933121.1 - - 467 119

View Full Product Details