Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HGD
Entrez Gene ID
3081
Gene Name
homogentisate 1,2-dioxygenase
Gene Aliases
AKU, HGO
Chromosome Location
Chr.3: 120628168 - 120682571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000187.3 NP_000178.2 2 - 3 546 161
XM_005247412.1 XP_005247469.1 2 - 3 546 161
XM_005247413.1 XP_005247470.1 2 - 3 546 161
XM_005247414.4 XP_005247471.1 2 - 3 546 161
XM_011512746.1 XP_011511048.1 2 - 3 546 161
XM_017006277.1 XP_016861766.1 2 - 3 159 161
GenBank mRNA AF045167.1 - 2 - 3 253 161
AK290782.1 - 2 - 3 252 161
AK313563.1 - 2 - 3 530 161
BC020792.1 - 2 - 3 252 161
BC071757.1 - 2 - 3 184 161
U63008.1 - 2 - 3 215 161
Z75048.1 - 2 - 3 215 161

View Full Product Details