The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004204.3 | NP_004195.2 | 5 - 6 | 1200 | 79 |
NM_148920.2 | NP_683721.1 | 5 - 6 | 1200 | 79 | |
GenBank mRNA | AB003723.1 | - | 5 - 6 | 1164 | 79 |
AF030177.1 | - | 5 - 6 | 1133 | 79 | |
AK300297.1 | - | 5 - 6 | 1167 | 79 | |
AK314361.1 | - | 6 - 7 | 1229 | 79 | |
BC006377.2 | - | 5 - 6 | 1150 | 79 |