The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174084.1 | NP_001167555.1 | 5 - 6 | 1384 | 80 |
NM_013274.3 | NP_037406.1 | 5 - 6 | 1321 | 80 | |
NR_033406.1 | 5 - 6 | 920 | 80 | ||
XM_011539650.1 | XP_011537952.1 | 5 - 6 | 989 | 80 | |
XM_011539651.1 | XP_011537953.1 | 5 - 6 | 989 | 80 | |
XM_011539652.1 | XP_011537954.1 | 5 - 6 | 1277 | 80 | |
XM_011539653.1 | XP_011537955.1 | 5 - 6 | 1270 | 80 | |
XM_011539657.1 | XP_011537959.1 | 5 - 6 | 1216 | 80 | |
XM_011539659.1 | XP_011537961.1 | 2 - 3 | 1265 | 80 | |
XM_011539660.1 | XP_011537962.1 | 3 - 4 | 521 | 80 | |
XM_017016089.1 | XP_016871578.1 | 3 - 4 | 794 | 80 | |
XM_017016091.1 | XP_016871580.1 | 2 - 3 | 1265 | 80 | |
GenBank mRNA | AF161019.1 | - | 4 - 5 | 950 | 80 |
AF283478.1 | - | 4 - 5 | 889 | 80 | |
AJ131890.1 | - | 5 - 6 | 1260 | 80 | |
AK021600.1 | - | 5 - 6 | 992 | 80 | |
AK022476.1 | - | 5 - 6 | 898 | 80 | |
AK293657.1 | - | 3 - 4 | 663 | 80 |