Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS51
Entrez Gene ID
738
Gene Name
VPS51, GARP complex subunit
Gene Aliases
ANG2, ANG3, C11orf2, C11orf3, FFR
Chromosome Location
Chr.11: 65096115 - 65111860 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013265.3 NP_037397.2 3 - 4 642 69
NR_073519.1 3 - 4 642 69
GenBank mRNA AF024631.2 - 3 - 4 546 69
AL833818.1 - 2 - 3 182 69
BC006555.2 - 2 - 3 288 69
BC007198.1 - 3 - 4 515 69
BC010540.1 - 3 - 4 521 69
BC010872.2 - 3 - 4 517 69
BC017438.1 - 3 - 4 515 69
BG391473.1 - 3 - 4 528 69
BI828867.1 - 3 - 4 569 69
CD388131.1 - 3 - 4 566 69

View Full Product Details