Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FTO
Entrez Gene ID
79068
Gene Name
fat mass and obesity associated
Gene Aliases
ALKBH9, BMIQ14, GDFD
Chromosome Location
Chr.16: 53703963 - 54114467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080432.2 NP_001073901.1 3 - 4 970 74
XM_017023655.1 XP_016879144.1 3 - 4 934 74
XM_017023656.1 XP_016879145.1 3 - 4 934 74
XM_017023657.1 XP_016879146.1 3 - 4 934 74
GenBank mRNA AB051539.1 - 3 - 4 754 74
AK096554.1 - 3 - 4 777 74

View Full Product Details