The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306147.1 | NP_001293076.1 | 8 - 9 | 1188 | 85 |
NM_018243.3 | NP_060713.1 | 6 - 7 | 968 | 85 | |
XM_006714259.3 | XP_006714322.1 | 6 - 7 | 834 | 85 | |
XM_017008400.1 | XP_016863889.1 | 7 - 8 | 977 | 85 | |
XM_017008401.1 | XP_016863890.1 | 7 - 8 | 989 | 85 | |
XM_017008402.1 | XP_016863891.1 | 7 - 8 | 989 | 85 | |
XM_017008403.1 | XP_016863892.1 | 6 - 7 | 929 | 85 | |
XM_017008404.1 | XP_016863893.1 | 6 - 7 | 929 | 85 | |
GenBank mRNA | AK001711.1 | - | 6 - 7 | 871 | 85 |
AK027279.1 | - | 4 - 5 | 545 | 85 | |
AK027633.1 | - | 3 - 4 | 383 | 85 | |
AK094473.1 | - | - | 191 | 85 | |
AK302700.1 | - | 8 - 9 | 1065 | 85 | |
AL110300.1 | - | 6 - 7 | 850 | 85 | |
BC008083.2 | - | 6 - 7 | 954 | 85 | |
BC063615.1 | - | 6 - 7 | 819 | 85 | |
GU727629.1 | - | 6 - 7 | 780 | 85 |