Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
Chromosome Location
Chr.10: 988409 - 1017768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012341.2 NP_036473.2 16 - 17 1819 89
GenBank mRNA AF120334.1 - 16 - 17 1771 86
AF325353.1 - 16 - 17 1748 89
AK001548.1 - 16 - 17 1766 89
AK001552.1 - 16 - 17 1821 89
AK222861.1 - 16 - 17 1766 89
AK295231.1 - 15 - 16 1662 89
AK301721.1 - 16 - 17 1934 89
AK302219.1 - 15 - 16 1624 89
BC033784.1 - 16 - 17 1742 89
BC038975.2 - 16 - 17 1780 89

View Full Product Details