The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079839.2 | NP_001073308.1 | 2 - 3 | 340 | 71 |
NM_001079840.2 | NP_001073309.1 | 2 - 3 | 277 | 71 | |
NM_001079841.2 | NP_001073310.1 | 2 - 3 | 277 | 71 | |
NM_001079842.2 | NP_001073311.2 | 2 - 3 | 340 | 71 | |
NM_001168254.1 | NP_001161726.1 | 2 - 3 | 296 | 71 | |
NM_017830.3 | NP_060300.1 | 2 - 3 | 277 | 71 | |
XM_017008329.1 | XP_016863818.1 | 2 - 3 | 483 | 71 | |
GenBank mRNA | AF251296.1 | - | 2 - 3 | 245 | 71 |
AF323665.1 | - | 2 - 3 | 255 | 71 | |
AF324350.1 | - | 2 - 3 | 255 | 71 | |
AK000462.1 | - | 2 - 3 | 232 | 71 | |
AL539411.3 | - | 2 - 3 | 212 | 71 | |
AL555289.3 | - | 2 - 3 | 170 | 71 | |
BC003409.1 | - | 2 - 3 | 225 | 71 | |
BC088361.1 | - | 2 - 3 | 234 | 71 | |
BI549536.1 | - | 3 - 4 | 389 | 71 | |
BM819316.1 | - | 2 - 3 | 126 | 71 | |
CD515566.1 | - | 2 - 3 | 239 | 71 | |
DA037995.1 | - | 2 - 3 | 277 | 71 | |
DT215975.1 | - | 1 - 2 | 81 | 71 |