Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OCIAD1
Entrez Gene ID
54940
Gene Name
OCIA domain containing 1
Gene Aliases
ASRIJ, OCIA, TPA018
Chromosome Location
Chr.4: 48831043 - 48861817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079839.2 NP_001073308.1 8 - 9 981 114
NM_001168254.1 NP_001161726.1 8 - 9 937 114
NM_017830.3 NP_060300.1 8 - 9 918 114
GenBank mRNA AF251296.1 - 8 - 9 886 114
AF323665.1 - 8 - 9 896 114
AK000462.1 - 8 - 9 873 114
AK123529.1 - 1 - 2 1400 114
BC003409.1 - 8 - 9 866 114
BC088361.1 - 8 - 9 875 114

View Full Product Details