Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C12orf50
Entrez Gene ID
160419
Gene Name
chromosome 12 open reading frame 50
Gene Aliases
-
Chromosome Location
Chr.12: 87980039 - 88029399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152589.1 NP_689802.1 5 - 6 538 130
XM_011537985.1 XP_011536287.1 5 - 6 781 130
XM_017018886.1 XP_016874375.1 4 - 5 478 130
XM_017018887.1 XP_016874376.1 5 - 6 781 130
XM_017018888.1 XP_016874377.1 4 - 5 478 130
GenBank mRNA AK093140.1 - 5 - 6 538 130
AL833338.1 - 5 - 6 781 130
BC038413.2 - 4 - 5 479 130

View Full Product Details