The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018402.1 | NP_060872.1 | 1 - 2 | 207 | 158 |
GenBank mRNA | AJ251549.1 | - | 1 - 2 | 207 | 158 |
BC066270.1 | - | 1 - 2 | 206 | 158 | |
BC066271.1 | - | 1 - 2 | 206 | 158 |